Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Eugenol, 99.6% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015991 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5991 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5991 Supplier MilliporeSigma Supelco Supplier No. 35995250MG
Add to Cart
Add to Cart

Eugenol is a phenolic compound and a major constituent of clove oil, which can find applications as an anti H. pylori agents and anti-bacterial agents. It can also act like a fungicidal agent, which can completely inhibit the growth of Saccharomyces cerevisiae, a species of yeast.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 97-53-0
Density 1.067 g/mL (at 25°C)
Flash Point 112°C - Closed Cup
Physical Form Neat
Linear Formula 4-(H2 C=CHCH2)C6H3-2-(OCH3)OH
MDL Number MFCD00008654
Melting Point -12°C to -10°C
Boiling Point 254°C
Synonym 2-Methoxy-4-(2-propenyl)phenol; 4-Allyl-2-methoxyphenol; 4-Allylguaiacol
Type Analytical Standard
Quantity 250 mg
Formula Weight 164.20
Percent Purity 99.6% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.