Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fenamidone, MilliporeSigma™ Supelco™

Catalog No. 111019825 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9825 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9825 Supplier MilliporeSigma Supelco Supplier No. 33965100MGR
Add to Cart
Add to Cart

Fenamidone is an imidazolinone fungicide. It is effective in treating downy mildew (Plasmopara viticola) in grapevines.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 161326-34-7
Physical Form Neat
Linear Formula C17H17N3 OS
MDL Number MFCD03792823
Synonym (S)-1-Anilino-4-methyl-2-methylthio-4-phenyl-2-imidazolin-5-one
Type Analytical Standard
Quantity 100 mg
Formula Weight 311.40
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.