Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fenbuconazol, MilliporeSigma™ Supelco™

Catalog No. 111019865 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9865 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9865 Supplier MilliporeSigma Supelco Supplier No. 31654100MG
Add to Cart
Add to Cart

Fenbuconazol is a triazole fungicide intended to control leaf spot, yellow and brown rust, powdery mildew and net blotch on wheat and barley. The mode of action of fenbuconazol involves inhibition of biosynthesis of sterol (a component of fungal cell membrane).

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 114369-43-6
Physical Form Neat
Linear Formula C19H17 ClN4
MDL Number MFCD01632336
Type Analytical Standard
Quantity 100 mg
Formula Weight 336.82
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.