Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fenoxaprop-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111019730 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9730 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9730 Supplier MilliporeSigma Supelco Supplier No. 45518250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenoxaprop-ethyl is a postemergence herbicide used for the control of annual and perennial weeds in crops. Its mode of action involves inhibiting the fatty acid biosynthesis in plant meristems, which in turn affects the activity of acetyl-coenzyme A carboxylase.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 66441-23-4
Physical Form Neat
Linear Formula C18H16 ClNO5
MDL Number MFCD01574785
Synonym Ethyl 2-{4-[(6-chlorobenzoxazol-2-yl)oxy]phenoxy}propionate
Type Analytical Standard
Quantity 250 mg
Formula Weight 361.78
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.