Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fenthion oxon sulfoxide, MilliporeSigma™ Supelco™

Catalog No. 111018116 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8116 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8116 Supplier MilliporeSigma Supelco Supplier No. 3237425MG
Add to Cart
Add to Cart

Fenthion oxon sulfoxide is a metabolite of fenthion, obtained by oxidative desulfuration. It is widely used as a pesticide in controlling insects in numerous crops. Its mode of action involves the inhibition of acetylcholinesterase.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 6552-13-2
Physical Form Neat
Linear Formula C10H15O5 PS
MDL Number MFCD01632380
Synonym Dimethyl 3-methyl-4-(methylsulfinyl)phenyl phosphate; Fenoxon sulfoxide
Type Analytical Standard
Quantity 25 mg
Formula Weight 278.26
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.