Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fenthion-sulfone, MilliporeSigma™ Supelco™

Catalog No. 111018642 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8642 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8642 Supplier MilliporeSigma Supelco Supplier No. 4602310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenthion-sulfone is one of the metabolites of fenthion which is classified under the organophosphorus group of pesticides.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 3761-42-0
Physical Form Neat
Linear Formula C10H15O5 PS2
MDL Number MFCD00155407
Type Analytical Standard
Quantity 10 mg
Formula Weight 310.33
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.