Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Fipronil carboxamide, MilliporeSigma™ Supelco™

Catalog No. 111018107 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8107 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8107 Supplier MilliporeSigma Supelco Supplier No. 3451910MG
Add to Cart
Add to Cart

Find more information here fipronil

TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 205650-69-7
Linear Formula C12H6 Cl2F6N4O2 S
MDL Number MFCD08273873
Synonym 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1 H-pyrazole-3-carboxamide
Type Analytical Standard
Quantity 10 mg
Formula Weight 455.16
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.