Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Flonicamid Metabolite TFNG, MilliporeSigma™ Supelco™

Catalog No. 111013504 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3504 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3504 Supplier MilliporeSigma Supelco Supplier No. 3456625MG
Add to Cart
Add to Cart

Flonicamid Metabolite TFNG is one of the metabolites of flonicamid, which is a selective aphicide.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 207502-65-6
Linear Formula C9H7F3N2O3
MDL Number MFCD11112215
Synonym N-[[4-(Trifluoromethyl)-3-pyridinyl]carbonyl]glycine; 4-(Trifluoromethyl)nicotinoyl glycine
Type Analytical Standard
Quantity 25 mg
Formula Weight 248.16
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.