Learn More
Description
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 69335-90-6 |
| Physical Form | Neat |
| Linear Formula | C16H14F3 NO4 |
| MDL Number | MFCD04112611 |
| Melting Point | 75°C to 80°C |
| Synonym | Methyl 2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 341.28 |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.