Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Flubendiamide, MilliporeSigma™ Supelco™

Catalog No. 111018684 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8684 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8684 Supplier MilliporeSigma Supelco Supplier No. 32801100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flubendiamide is a novel insecticide that is grouped under the family of phthalic acid diamides. It is largely used against lepidopteron pests in various annual and perennial crops.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 272451-65-7
Physical Form Neat
Linear Formula C23H22F7 IN2O4 S
MDL Number MFCD09953911
Synonym N2-[1,1-Dimethyl-2-(methylsulfonyl)ethyl]-3-iodo-N1-{2-methyl-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]phenyl}-1,2-benzenedicarboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 682.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.