Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Flucycloxuron, Mixture of (E) and (Z) isomers, MilliporeSigma™ Supelco™

Catalog No. 111012627 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2627 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2627 Supplier MilliporeSigma Supelco Supplier No. 3252925MG
Add to Cart
Add to Cart

Flucycloxuron is a new derivative of benzoylphenylurea (BPU) that acts against mites and insects by inhibiting the chitin synthesis that results in abnormal endocuticular deposition and abortive molting in the target species.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 113036-88-7
Physical Form Neat
Linear Formula C25H20 ClF2N3O3
MDL Number MFCD01632755
Synonym N-[[[4-[[[[(4-Chlorophenyl)cyclopropylmethylene]amino]oxy]methyl]phenyl]amino]carbonyl]-2,6-difluorobenzamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 483.89
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.