Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Flufenoxuron, MilliporeSigma™ Supelco™

Catalog No. 111012288 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2288 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2288 Supplier MilliporeSigma Supelco Supplier No. 31594250MG
Add to Cart
Add to Cart

Flufenoxuron is an acylurea compound mostly used as an insecticide. It inhibits chitin synthesis in pests making it a slow acting growth regulator.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 101463-69-8
Physical Form Neat
Linear Formula C21H11 ClF6N2O3
MDL Number MFCD00274597
Synonym N-{4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenylaminocarbonyl}-2,6-difluoro-benzamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 488.77
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.