Learn More
PESTANAL™ Flumiclorac, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Product Line | PESTANAL |
| CAS | 87547-04-4 |
| Linear Formula | C16H13 ClFNO5 |
| MDL Number | MFCD18449905 |
| Synonym | 2-Chloro-4-fluoro-5-(3,4,5,6-tetrahydrophthalimido)phenoxyacetic acid; 2-[2-Chloro-4-fluoro-5-(1,3,4,5,6,7-hexahydro-1,3-dioxo-2 H-isoindol-2-yl)phenoxy]acetic acid; [2-Chloro-5-(cyclohex-1-ene-1,2-dicarboximido)-4-fluororophenoxy]acetic acid |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 353.73 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.