Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Flupyradifurone, MilliporeSigma™ Supelco™

Catalog No. 111015956 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5956 Supplier MilliporeSigma Supelco Supplier No. 37050100MG
Add to Cart
Add to Cart

Flupyradifurone is a systemic insecticide, that belongs to the butenolide class of insecticides, which can be used for agricultural purposes in order to control a broad spectrum of sucking insects.

TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 951659-40-8
Physical Form Neat
Linear Formula C12H11 ClF2N2O2
MDL Number MFCD22373627
Synonym 4-[(6-Chloro-3-pyridylmethyl)(2,2-difluoroethyl)amino]furan-2(5 H)-one
Type Analytical Standard
Quantity 100 mg
Formula Weight 288.68
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.