Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Foramsulfuron, MilliporeSigma™ Supelco™

Catalog No. 111013339 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3339 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3339 Supplier MilliporeSigma Supelco Supplier No. 33977100MGR
Add to Cart
Add to Cart

Foramsulfuron is a sulfonylurea herbicide used in controlling a wide spectrum of weeds. Its mode of action involves inhibiting the enzyme activity of acetolactate synthase (ALS) which is involved in biosynthesis of branched amino acids.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 173159-57-4
Physical Form Neat
Linear Formula C17H20N6O7 S
MDL Number MFCD04972564
Synonym 2-[3-(4,6-Dimethoxy-2-pyrimidinyl)ureidosulfonyl]-4-(formamido)-N,N-dimethylbenzamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 452.44
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.