Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Formothion Solution, ∼80 in Xylene, MilliporeSigma™ Supelco™

Catalog No. 111018733 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8733 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8733 Supplier MilliporeSigma Supelco Supplier No. 46424250MG
Add to Cart
Add to Cart

Formothion is classified under the group of organophosphorus (OP) insecticides utilized for the protection of crops from a variety of sucking pests and some biting and chewing insects. Its mode of action involves the inhibition of cholinesterase enzyme and it is a suspected endocrine disruptor.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2540-82-1
Flash Point 25°C
Physical Form Neat
Linear Formula C6H12 NO4 PS2
MDL Number MFCD00210289
Synonym S-[2-(Formylmethylamino)-2-oxoethyl] O,O-dimethyl dithiophosphate
Type Analytical Standard
Quantity 250 mg
Formula Weight 257.27
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 80% in xylene
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.