Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ β-HCH, MilliporeSigma™ Supelco™

Catalog No. 111019300 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9300 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9300 Supplier MilliporeSigma Supelco Supplier No. 33376100MG
Add to Cart
Add to Cart

β-Hexachlorocyclohexane is an isomer of hexachlorocyclohexane. It is an organochlorine pesticide with carcinogenic effects on both humans and animals.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 319-85-7
Physical Form Neat
Linear Formula C6H6 Cl6
MDL Number MFCD00135948
Melting Point ≥300°C
Synonym beta-1,2,3,4,5,6-Hexachlorocyclohexane
Type Analytical Standard
Quantity 100 mg
Formula Weight 290.83
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.