Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Imazapyr, MilliporeSigma™ Supelco™

Catalog No. 111015608 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5608 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5608 Supplier MilliporeSigma Supelco Supplier No. 37877100MG
Add to Cart
Add to Cart

Imazapyr is a broad spectrum non-selective herbicide, belonging to the family of imidazolinones, which can find applications in weed control management in cereals and sugar beet. Its mode of action involves the inhibition of the synthesis of branched-chain amino acids in plants.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 81334-34-1
Physical Form Neat
Linear Formula C13H15N3O3
MDL Number MFCD00144470
Type Analytical Standard
Quantity 100 mg
Formula Weight 261.28
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.