Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Imidacloprid, MilliporeSigma™ Supelco™

Catalog No. 111019462 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9462 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9462 Supplier MilliporeSigma Supelco Supplier No. 37894100MG
Add to Cart
Add to Cart

Imidacloprid is a systemic chloronicotinyl insecticide, which can be effectively used in agriculture to control pests like aphids, scale insects, whiteflies, leafhoppers, some coleoptera and some lepidoptera species.Find more information here: Neonicotinoids

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 138261-41-3
Physical Form Neat
Linear Formula C9H10 ClN5O2
MDL Number MFCD00468059
Melting Point 140°C to 146°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 255.66
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.