Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Imidacloprid-olefin, MilliporeSigma™ Supelco™

Catalog No. 111012646 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2646 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2646 Supplier MilliporeSigma Supelco Supplier No. 3453410MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imidacloprid-olefin is a metabolite of the insecticide, imidacloprid.Find more information here: Neonicotinoids

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 115086-54-9
Linear Formula C9H8 ClN5O2
MDL Number MFCD02660798
Synonym 1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-1 H-imidazol-2-amine
Type Analytical Standard
Quantity 10 mg
Formula Weight 253.65
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.