Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Imiprothrin, MilliporeSigma™ Supelco™

Catalog No. 111018006 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8006 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8006 Supplier MilliporeSigma Supelco Supplier No. 3344250MG
Add to Cart
Add to Cart

Imiprothrin is a kind of pyrethroid consisting of an imidazolydinyl ring, which can find applications as a potent knock-down insecticide.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 72963-72-5
Flash Point 110°C
Physical Form Neat
Linear Formula C17H22N2O4
MDL Number MFCD03703407
Synonym 2-Propargylsuccinimidylmethyl chrysanthemate
Type Analytical Standard
Quantity 50 mg
Formula Weight 318.37
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.