Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Indoxacarb, MilliporeSigma™ Supelco™

Catalog No. 111019251 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9251 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9251 Supplier MilliporeSigma Supelco Supplier No. 3396925MGR
Add to Cart
Add to Cart

Indoxacarb belongs to oxadiazine class of insecticides, showing broad-spectrum activity against Lepidoptera and other pests. Its mode of action involves blocking of voltage-gated sodium channels and binding to neuronal nicotinic acetylcholine receptors.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 144171-61-9
Physical Form Neat
Linear Formula C22H17 ClF3N3O7
MDL Number MFCD03792834
Synonym Methyl 7-chloro-2,5-dihydro-2-[N-(methoxycarbonyl)-4-(trifluoromethoxy)anilinocarbonyl]indeno[1,2-e][1,3,4 ]oxadiazine-4 A(3 H)carboxylate
Type Analytical Standard
Quantity 25 mg
Formula Weight 527.83
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.