Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Isoproturon-d6, MilliporeSigma™ Supelco™

Catalog No. 111018737 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8737 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8737 Supplier MilliporeSigma Supelco Supplier No. 3401710MGR
Add to Cart
Add to Cart

Isoproturon-d6 is an isotopically labeled standard of the phenylurea herbicide isoproturon, which is used for the pre- or post-emergence control of weeds in fruits, cotton and other crops.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 217487-17-7
Physical Form Neat
Linear Formula C12H12D6N2 O
MDL Number MFCD01632296
Synonym 3-(4-Isopropylphenyl)-1,1-dimethylurea-d6
Type Analytical Standard
Quantity 10 mg
Formula Weight 212.32
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.