Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Isoxadifen-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111018520 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8520 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-8520 Supplier MilliporeSigma Supelco Supplier No. 33799100MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isoxadifen-ethyl can play the role as a safener with pesticide fenoxaprop-ethyl, which can be an effective combination in agriculture to protect rice. It can also be used as a herbicide for cereal crops to combat grassy weeds.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 163520-33-0
Physical Form Neat
Linear Formula C18H17 NO3
MDL Number MFCD03792846
Synonym Ethyl 4,5-dihydro-5,5-diphenylisoxazol-3-carboxylate
Type Analytical Standard
Quantity 100 mg
Formula Weight 295.33
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.