Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Isoxaflutole, MilliporeSigma™ Supelco™

Catalog No. 111018879 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8879 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8879 Supplier MilliporeSigma Supelco Supplier No. 46437100MG
Add to Cart
Add to Cart

Isoxaflutole is a novel pre-emergence herbicide widely employed for the protection of corn and sugarcane against broadleaf and grass weeds. Its mode of action involves the facilitation of characteristic foliar bleaching activity in target species preceding the plant death.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 141112-29-0
Flash Point >130°C
Physical Form Neat
Linear Formula C15H12F3 NO4 S
MDL Number MFCD03095707
Synonym 5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole
Type Analytical Standard
Quantity 100 mg
Formula Weight 359.32
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.