Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Malaoxon, MilliporeSigma™ Supelco™

Catalog No. 111019296 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9296 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9296 Supplier MilliporeSigma Supelco Supplier No. 36142100MG
Add to Cart
Add to Cart

Malaoxon is one of the major metabolites of malathion and a widely used insecticide in agriculture. Its mode of action includes DNA damage in human lymphocytes by the mechanism of oxidative action.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1634-78-2
Flash Point >100°C
Physical Form Neat
Linear Formula C10H19O7 PS
MDL Number MFCD00055461
Type Analytical Standard
Quantity 100 mg
Formula Weight 314.29
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.