Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Malathion, MilliporeSigma™ Supelco™

Catalog No. 111020185 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0185 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0185 Supplier MilliporeSigma Supelco Supplier No. 36143100MG
Add to Cart
Add to Cart

Malathion is a commonly used organophosphate pesticide, which is generally sprayed on aquatic habitats in order to have a control over mosquitoes that carry malaria and the west nile virus.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 121-75-5
Physical Form Neat
Linear Formula C10H19O6 PS2
MDL Number MFCD00036233
Type Analytical Standard
Quantity 100 mg
Formula Weight 330.36
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.