Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ MCPA-2-ethylhexyl ester, MilliporeSigma™ Supelco™

Catalog No. 111019354 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9354 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9354 Supplier MilliporeSigma Supelco Supplier No. 33394100MG
Add to Cart
Add to Cart

MCPA-2-ethylhexyl ester is a systemic, post-emergent herbicide used for the selective control of annual and perennial broad leaved weeds in cereals and other specific crops.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 29450-45-1
Physical Form Neat
Linear Formula C17H25 ClO3
MDL Number MFCD00144729
Synonym 2-Ethylhexyl (4-chloro-2-methylphenoxy)acetate
Type Analytical Standard
Quantity 100 mg
Formula Weight 312.83
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.