Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ MCPA-thioethyl, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019192 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9192 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9192 Supplier MilliporeSigma Supelco Supplier No. 9454925MG

MCPA-thioethyl is a selective, systemic phenoxyacetic herbicide used for the postemergence control of annual and perennial broadleaved weeds in cereals. Its mode of action involves the inhibition of growth in meristematic region.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line PESTANAL
CAS 25319-90-8
Physical Form Neat
Linear Formula C11H13 ClO2 S
MDL Number MFCD01671920
Synonym S-Ethyl 4-chloro-o-tolyloxythioacetate; Phenothiol
Type Analytical Standard
Quantity 25 mg
Formula Weight 244.74
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.