Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Mepiquat iodide-(methyl-d3), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013922 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3922 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3922 Supplier MilliporeSigma Supelco Supplier No. 7827810MG
Add to Cart
Add to Cart

Mepiquat iodide-(methyl-d3) is an iodide salt of mepiquat which is widely used as a plant growth regulator in agriculture.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 32317-85-4
Linear Formula C7D3H13 in.
MDL Number MFCD00032083
Synonym 1-Methyl-1-methyl-d3-piperidinium iodide
Type Analytical Standard
Quantity 10 mg
Formula Weight 244.13
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.