Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Metamitron, MilliporeSigma™ Supelco™

Catalog No. 111019807 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9807 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9807 Supplier MilliporeSigma Supelco Supplier No. 36154100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metamitron is a 1,2,4-triazine herbicide, which is yellow crystalline in nature and can easily degrade in soil to form stable metabolites. It can be prepared via reaction of N-acetyl-benzylhydrazone with hydroxylamine in the presence of potassium hydroxide in pyridine.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 41394-05-2
Physical Form Neat
Linear Formula C10H10N4 O
MDL Number MFCD00055524
Melting Point 165°C to 170°C
Synonym 4-Amino-4,5-dihydro-3-methyl-6-phenyl-1,2,4-triazin-5-one
Type Analytical Standard
Quantity 100 mg
Formula Weight 202.21
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.