Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Methiocarb sulfoxide, MilliporeSigma™ Supelco™

Catalog No. 111019260 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9260 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9260 Supplier MilliporeSigma Supelco Supplier No. 34177100MG
Add to Cart
Add to Cart

Methiocarb sulfoxide, is a carbamate insecticide, which is used to control rice insects. It is a metabolite of methiocarb present in honey.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2635-10-1
Physical Form Neat
Linear Formula C11H15 NO3 S
MDL Number MFCD00155393
Synonym 4-Methylsulfinyl-3,5-xylyl-N-methylcarbamate; Mercaptodimethur sulfoxide; Mesurol sulfoxide; Methiocarb breakdown product
Type Analytical Standard
Quantity 100 mg
Formula Weight 241.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.