Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Methyl Paraben, MilliporeSigma™ Supelco™

Catalog No. 111016026 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6026 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6026 Supplier MilliporeSigma Supelco Supplier No. 47889
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Paraben is a stable, non-volatile, methyl ester of p-hydroxybenzoic acid. It can find applications as a broad-spectrum antimicrobial preservative in foods, drugs and cosmetics. It is found to be noncarcinogenic and is highly effective against fungi like molds and yeast as well as bacteria.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line PESTANAL
CAS 99-76-3
Physical Form Neat
Linear Formula HOC6H4 CO2 CH3
MDL Number MFCD00002352
Melting Point 125°C to 128°C
Synonym Methyl 4-hydroxybenzoate; p-Hydroxybenzoic acid methyl ester; Methyl paraben; NIPAGIN
Type Food Additive Standard
Quantity 1000 mg
Formula Weight 152.15
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.