Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Metolachlor, predominantly S-Metolachlor, ∼60% (S-Metolachlor (HPLC-ER)), MilliporeSigma™ Supelco™

Catalog No. 111020441 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0441 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0441 Supplier MilliporeSigma Supelco Supplier No. 33859100MGR
Add to Cart
Add to Cart

S-Metolachlor is a herbicide and an isomer of metolachlor, which can be used for selective grass weed control, and it can reduce the chemical load to the environment likewise give good efficiency.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 87392-12-9
Physical Form Neat
Linear Formula C15H22 ClNO2
MDL Number MFCD04967118
Synonym 2-Chloro-N-(2-ethyl-6-methylphenyl)-N-[(1 S)-2-methoxy-1-methylethyl]acetamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 283.79
Percent Purity 60% (S to Metolachlor (HPLC to ER))
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.