Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Nicosulfuron, MilliporeSigma™ Supelco™

Catalog No. 111019028 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9028 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9028 Supplier MilliporeSigma Supelco Supplier No. 34210100MGR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicosulfuron belongs to the group of sulfonylurea herbicides, which can be largely used to control grasses and broad leaved weed species in crop protection like in corn.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 111991-09-4
Physical Form Neat
Linear Formula C15H18N6O6 S
MDL Number MFCD00203303
Synonym 1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(dimethylcarbamoyl)-2-pyridylsulfonyl]urea; 2-[(4,6-Dimethoxypyrimidin-2-ylcarbamoyl)sulfamoyl]-N,N-dimethylnicotinamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 410.41
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.