Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Nitrapyrin, MilliporeSigma™ Supelco™

Catalog No. 111018959 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8959 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8959 Supplier MilliporeSigma Supelco Supplier No. 33372100MG
Add to Cart
Add to Cart

Nitrapyrin is a nitrification inhibitor, which can also undergo hydrolysis to form 6-chloropicolinic acid, that can inhibit methane oxidation by M. trichosporium OB3b.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1929-82-4
Flash Point 100°C - Closed Cup
Physical Form Neat
Linear Formula C6H3 Cl4 N
MDL Number MFCD00072500
Melting Point 61°C to 65°C
Synonym 2-Chloro-6-(trichloromethyl)pyridine; CP
Type Analytical Standard
Quantity 100 mg
Formula Weight 230.91
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.