Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PESTANAL™ Nitrobenzene, ≥99% (GC), MilliporeSigma™ Supelco™
Nitrobenzene is a carcinogenic compound which has been employed extensively in dyes, pesticides and explosives industries.
Supplier: MilliporeSigma Supelco 060845MLF
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
98-95-3 | |
Analytical Standard | |
C6H5 NO2 | |
123.11 | |
Analytical Standard | |
0.15 mmHg (20°C) | |
1.196 g/mL (at 25°C) | |
210°C to 211°C |
5 mL | |
PESTANAL | |
MFCD00007043 | |
≥99% (GC) | |
899°F | |
4.2 (vs. air) | |
5°C to 6°C | |
88°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
Spot an opportunity for improvement?
Provide Content Correction