Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Nitrobenzene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020664 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0664 1 mL
11-101-9829 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0664 Supplier MilliporeSigma Supelco Supplier No. 060841MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrobenzene is a carcinogenic compound which has been employed extensively in dyes, pesticides and explosives industries.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 98-95-3
Product Line PESTANAL
Ignition Point 899°F
Vapor Pressure 0.15 mmHg (20°C)
Vapor Density 4.2 (vs. air)
Density 1.196 g/mL (at 25°C)
Flash Point 88°C
Linear Formula C6H5 NO2
MDL Number MFCD00007043
Melting Point 5°C to 6°C
Boiling Point 210°C to 211°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 123.11
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.