Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Novaluron, MilliporeSigma™ Supelco™

Catalog No. 111018908 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8908 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8908 Supplier MilliporeSigma Supelco Supplier No. 3241925MG
Add to Cart
Add to Cart

Novaluron (Rimon) is a type of benzoylphenyl urea,3 that acts as an insect growth regulator by inhibiting chitin synthesis. It acts as a powerful toxicant for controlling lepidopteran larvae. It is suitable in the rainy and tropics seasons, as it is a rainfast compound.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 116714-46-6
Flash Point 202°C - Closed Cup
Physical Form Neat
Linear Formula C17H9 ClF8N2O4
MDL Number MFCD04112632
Synonym 1-[3-Chloro-4-(1,1,2-trifluoro-2-trifluoromethoxyethoxy)phenyl]-3-(2,6-difluorobenzoyl)urea
Type Analytical Standard
Quantity 25 mg
Formula Weight 492.70
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.