Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Paraoxon-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111019459 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9459 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9459 Supplier MilliporeSigma Supelco Supplier No. 36186100MG
Add to Cart
Add to Cart

Paraoxon-ethyl is an organophosphate pesticide and an inhibitor of acetylcholinesterase, which can also serve as a well-known surrogate of chemical warfare agents. It can undergo degradation by using titania nanoparticles, which are immobilized on glass or quartz substrates.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 311-45-5
Density 1.274 g/mL (at 25°C)
Physical Form Neat
Linear Formula O2 NC6H4 OP(O)(OC2H5)2
MDL Number MFCD00007316
Synonym O,O-Diethyl O-(4-nitrophenyl) phosphate; Diethyl p-nitrophenyl phosphate; Paraoxon; Paraoxon-ethyl
Type Analytical Standard
Quantity 100 mg
Formula Weight 275.20
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.