Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Parathion-methyl-d6 (dimethyl-d6), ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018027 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8027 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8027 Supplier MilliporeSigma Supelco Supplier No. 940935MG
Add to Cart
Add to Cart

Specifications

CAS 96740-32-8
Product Line PESTANAL
Purity Grade Notes identification (LC-MS): identity conforms
Linear Formula C8D6H4 NO5 PS
MDL Number MFCD03428104
Synonym O,O-Dimethyl-d6 O-(4-nitrophenyl) phosphorothioate; Methyl parathion-d6
Type Analytical Standard
Quantity 5 mg
Formula Weight 269.24
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.