Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Parathion, MilliporeSigma™ Supelco™

Catalog No. 111020281 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0281 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0281 Supplier MilliporeSigma Supelco Supplier No. 45607100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 56-38-2
Flash Point 120°C
Physical Form Neat
Linear Formula C10H14 NO5 PS
MDL Number MFCD00036219
Synonym O,O-Diethyl O-(4-nitrophenyl) phosphorothioate; Ethylparathione; Parathion-ethyl
Type Analytical Standard
Quantity 100 mg
Formula Weight 291.26
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.