Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Permethrin-(phenoxy-d5) Mixture of cis and trans isomers, ≥98.0% (sum of isomers, HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018763 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8763 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8763 Supplier MilliporeSigma Supelco Supplier No. 745675MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Line PESTANAL
CAS 1794760-19-2
Linear Formula C21D5H15 Cl2O3
Synonym Permethrin-d5; [3-(Phenoxy-d5)phenyl]methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate
Type Analytical Standard
Quantity 5 mg
Formula Weight 396.32
Percent Purity ≥98.0% (sum of isomers, HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.