Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Phorate, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017273 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7273 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7273 Supplier MilliporeSigma Supelco Supplier No. 33388100MG
Add to Cart
Add to Cart

Phorate is an organophosphate insecticide used to control sap-feeding insects including various beetles, mites, grubs, and worms in agricultural crop cultivation.

TRUSTED_SUSTAINABILITY

Specifications

CAS 298-02-2
Product Line PESTANAL
Physical Form Neat
Linear Formula C7H17O2PS3
MDL Number MFCD00036348
Synonym O,O-Diethyl S-(ethylthiomethyl) phosphorodithioate
Quantity 100 mg
Formula Weight 260.38
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.