Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Picloram, MilliporeSigma™ Supelco™

Catalog No. 111013415 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3415 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3415 Supplier MilliporeSigma Supelco Supplier No. 36774250MGR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard for agricultural and environmental applications.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Picloram
CAS 2-1-1918
Product Line PESTANAL
Physical Form Neat
Linear Formula C6H3 Cl3N2O2
MDL Number MFCD00012101
Melting Point 200°C
Synonym 4-Amino-3,5,6-trichloropyridine-2-carboxylic acid; 4-Amino-3,5,6-trichloropicolinic acid; Picloram
Type Analytical Standard
Industry Type Environmental Testing
Molecular Weight (g/mol) 241.46
Quantity 250 mg
Formula Weight 241.46
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.