Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Picolinafen, MilliporeSigma™ Supelco™

Catalog No. 111019351 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9351 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9351 Supplier MilliporeSigma Supelco Supplier No. 37912100MGR
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 137641-05-5
Physical Form Neat
Linear Formula C19H12F4N2O2
MDL Number MFCD04040061
Synonym N-(4-Fluorophenyl)-6-(alpha,alpha,alpha-trifluoro-m-tolyloxy)picolinamide; N-(4-Fluorophenyl)-6-[3-(trifluoromethyl)phenoxy]-2-pyridinecarboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 376.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.