Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Pirimiphos-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111020465 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0465 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0465 Supplier MilliporeSigma Supelco Supplier No. 45628250MG
Add to Cart
Add to Cart

Pirimiphos-ethyl belongs to the class of organophosphorus pesticides, with contact and respiratory action and is commonly employed for the protection of crops including vegetables, bananas, turf and mushrooms. Its mode of action involves the inhibition of acetylcholinesterase activity.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 23505-41-1
Physical Form Neat
Linear Formula C13H24N3O3 PS
MDL Number MFCD00055295
Type Analytical Standard
Quantity 250 mg
Formula Weight 333.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.