Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Propaquizafop, MilliporeSigma™ Supelco™

Catalog No. 111012472 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2472 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2472 Supplier MilliporeSigma Supelco Supplier No. 31572250MG
Add to Cart
Add to Cart

Propaquizafop is an aryloxyphenoxypropionate herbicide primarily used for post emergence control of weeds in agricultural products. Its mode of action involves inhibition of the plastid acetyl coenzyme A carboxylase (ACCase), a key enzyme in fatty acid biosynthesis.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 111479-05-1
Physical Form Neat
Linear Formula C22H22 ClN3O5
MDL Number MFCD02181171
Type Analytical Standard
Quantity 250 mg
Formula Weight 443.88
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.