Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Propargite, technical Mixture, MilliporeSigma™ Supelco™

Catalog No. 111020030 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0030 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0030 Supplier MilliporeSigma Supelco Supplier No. 32051100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargite, a non-systemic acaricide, is most commonly used on crops such as cotton, vines, nuts, tea etc for controlling phytophagous mite infestation.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2312-35-8
Flash Point 72°C - Closed Cup
Physical Form Neat
Linear Formula C19H26O4 S
MDL Number MFCD00072448
Synonym 2-(4-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite
Type Analytical Standard
Quantity 100 mg
Formula Weight 350.47
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.