Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Propoxur, MilliporeSigma™ Supelco™

Catalog No. 111020223 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0223 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0223 Supplier MilliporeSigma Supelco Supplier No. 45644250MG
Add to Cart
Add to Cart

Propoxur is a non-systemic carbamate insecticide, mainly used for agricultural and non-agricultural applications, for controlling a variety of insects pests such as ants, crickets flies, mosquitoes, cockroaches, chewing and sucking insects.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 114-26-1
Physical Form Neat
Linear Formula C11H15 NO3
MDL Number MFCD00055455
Type Analytical Standard
Quantity 250 mg
Formula Weight 209.24
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.